Product Details

SNP ID
rs4692080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:26164101 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATGCTTCTGTTGGTAGGTGAT[A/T]CTGATCACGTTGGTTGTTTACATCT
Phenotype
MIM: 147183
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RBPJ PubMed Links

Gene Details

Gene
RBPJ
Gene Name
recombination signal binding protein for immunoglobulin kappa J region
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005349.3 Intron NP_005340.2
NM_015874.4 Intron NP_056958.3
NM_203283.2 Intron NP_976028.1
NM_203284.2 Intron NP_976029.1
XM_005248161.3 Intron XP_005248218.1
XM_011513840.2 Intron XP_011512142.1
XM_017008170.1 Intron XP_016863659.1
XM_017008171.1 Intron XP_016863660.1
XM_017008172.1 Intron XP_016863661.1
XM_017008173.1 Intron XP_016863662.1
XM_017008174.1 Intron XP_016863663.1
XM_017008175.1 Intron XP_016863664.1

View Full Product Details