Product Details

SNP ID
rs4547400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:62633189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATAATTTCTTAGCAAACAATA[C/G]TAAATTATTGCTATTTTTATTAGAA
Phenotype
MIM: 612264
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRC2 PubMed Links

Gene Details

Gene
MRC2
Gene Name
mannose receptor C type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006039.4 Intron NP_006030.2

View Full Product Details