Product Details

SNP ID
rs4759237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56011956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGATTTCAACATATTTTTGCTCA[C/G]ATTTCACTCAGTTCAGGAAGGCATA
Phenotype
MIM: 606239 MIM: 606887
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IKZF4 PubMed Links

Gene Details

Gene
IKZF4
Gene Name
IKAROS family zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022465.3 Intron NP_071910.3
Gene
LOC105369781
Gene Name
uncharacterized LOC105369781
There are no transcripts associated with this gene.

Gene
SUOX
Gene Name
sulfite oxidase
There are no transcripts associated with this gene.

View Full Product Details