Product Details

SNP ID
hCV28002267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47955890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACTTGGGGAATGGAAAAACAGA[A/G]GATCCAGCTCCTGAGACTATCATAA
Phenotype
MIM: 603287
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNPO PubMed Links

Gene Details

Gene
PNPO
Gene Name
pyridoxamine 5'-phosphate oxidase
There are no transcripts associated with this gene.

Gene
PRR15L
Gene Name
proline rich 15 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024320.3 Intron NP_077296.1

View Full Product Details