Product Details

SNP ID
rs4963770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:24567683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCTGTCTCGTCAAATGCAATGT[C/G]CATATCAAATTTTGAAAGTGAGCTT
Phenotype
MIM: 604975
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00477 PubMed Links

Gene Details

Gene
LINC00477
Gene Name
long intergenic non-protein coding RNA 477
There are no transcripts associated with this gene.

Gene
SOX5
Gene Name
SRY-box 5
There are no transcripts associated with this gene.

View Full Product Details