Product Details

SNP ID
rs4273596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132815435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGCCTGCACCTCCAGCTTCTTT[C/T]TGAAGCATGACCCATCTCCAGAGAT
Phenotype
MIM: 611179
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHROOM1 PubMed Links

Gene Details

Gene
SHROOM1
Gene Name
shroom family member 1
There are no transcripts associated with this gene.

Gene
SOWAHA
Gene Name
sosondowah ankyrin repeat domain family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175873.5 2112 UTR 3 NP_787069.4

View Full Product Details