Product Details

SNP ID
rs4356044
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:158492098 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGGTATACAGCCTCAGGAACAAC[G/T]ATGTGCAGATGGCTATTCGGAAACT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10R3P PubMed Links

Gene Details

Gene
OR10R3P
Gene Name
olfactory receptor family 10 subfamily R member 3 pseudogene
There are no transcripts associated with this gene.

View Full Product Details