Product Details

SNP ID
rs3944110
Assay Type
Functionally Tested
NCBI dbSNP Submissions
33
Location
Chr.1:35575330 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCACTTCCCTGTTTCCCTAAGAG[A/G]TGGGAATGGAAGTGCTGTTCCCACG
Phenotype
MIM: 608458 MIM: 614428
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NCDN PubMed Links

Gene Details

Gene
NCDN
Gene Name
neurochondrin
There are no transcripts associated with this gene.

Gene
TFAP2E
Gene Name
transcription factor AP-2 epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178548.3 Intron NP_848643.2
XM_017001139.1 Intron XP_016856628.1

View Full Product Details