Product Details

SNP ID
rs4693446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82484805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACCTTCACTGCCCCCATCTTGT[A/T]CCAGGCCTGATAGACCACGTCCCTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105377310 PubMed Links

Gene Details

Gene
LOC105377310
Gene Name
syncytin-1-like
There are no transcripts associated with this gene.

Gene
TMEM150C
Gene Name
transmembrane protein 150C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080506.1 1534 UTR 3 NP_001073975.1

View Full Product Details