Product Details

SNP ID
rs7813676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142198919 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGAGAACATTCTAGACTCACCTC[A/C]CACCTGCCCCCTCACTAACAGCCTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00051 PubMed Links

Gene Details

Gene
LINC00051
Gene Name
long intergenic non-protein coding RNA 51
There are no transcripts associated with this gene.

View Full Product Details