Product Details
- SNP ID
-
rs6527727
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:16794659 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGTATCAGGATTGGTCCATATGGA[G/T]CTAGGGAAGGATGACTTCATAAAAT
- Phenotype
-
MIM: 300677
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TXLNG
PubMed Links
Gene Details
- Gene
- TXLNG
- Gene Name
- taxilin gamma
View Full Product Details