Product Details
- SNP ID
-
rs1128272
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:68946438 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTGTACTTGACCCAGGTGTATTTG[C/T]TGGAGAACATGTTGTCCAGAGCCTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GKN2
PubMed Links
Gene Details
- Gene
- GKN2
- Gene Name
- gastrokine 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182536.2 |
447 |
Missense Mutation |
AAC,AGC |
N113S |
NP_872342.2 |
View Full Product Details