Product Details

SNP ID
rs8063977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2519456 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTGGAGGACTGCAGGGAGGGGGG[C/T]GGTTCACCTGTGGGTGGTGACCCGG
Phenotype
MIM: 108745 MIM: 613577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMDHD2 PubMed Links

Gene Details

Gene
AMDHD2
Gene Name
amidohydrolase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145815.1 Intron NP_001139287.1
NM_015944.3 Intron NP_057028.2
XM_017023262.1 Intron XP_016878751.1
XM_017023263.1 Intron XP_016878752.1
XM_017023264.1 Intron XP_016878753.1
XM_017023265.1 Intron XP_016878754.1
XM_017023266.1 Intron XP_016878755.1
XM_017023267.1 Intron XP_016878756.1
Gene
ATP6V0C
Gene Name
ATPase H+ transporting V0 subunit c
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198569.1 Intron NP_001185498.1
NM_001694.3 Intron NP_001685.1
Gene
TBC1D24
Gene Name
TBC1 domain family member 24
There are no transcripts associated with this gene.

View Full Product Details