Product Details
- SNP ID
-
hCV28980177
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:40915753 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTTTCACCCTTTCAGATATGCTA[A/G]TCAGCACAATAGGCTTAAACATTTT
- Phenotype
-
MIM: 217070
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C7
PubMed Links
Gene Details
- Gene
- C7
- Gene Name
- complement component 7
There are no transcripts associated with this gene.
View Full Product Details