Product Details

SNP ID
rs7586341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216032255 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTAAACTTGTAACTCAGTCCAGA[C/G]AAATGAAAACAAAAGCCAAATGGTC
Phenotype
MIM: 605843
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PECR PubMed Links

Gene Details

Gene
PECR
Gene Name
peroxisomal trans-2-enoyl-CoA reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018441.5 Intron NP_060911.2
XM_011511478.2 Intron XP_011509780.1

View Full Product Details