Product Details

SNP ID
rs8025748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:69082527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAATAGTATGAATTCATTTGCA[A/G]GCGTGCTCAGATAAGATAGAGTCTT
Phenotype
MIM: 616492
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EWSAT1 PubMed Links

Gene Details

Gene
EWSAT1
Gene Name
Ewing sarcoma associated transcript 1
There are no transcripts associated with this gene.

View Full Product Details