Product Details
- SNP ID
-
rs8033060
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:48991868 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAACAATTTTTTGGTCAAATTAGAG[C/T]AGAGGAGGTGGAGTCCTGAAGCAAA
- Phenotype
-
MIM: 615756
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SECISBP2L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73404249] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SECISBP2L
- Gene Name
- SECIS binding protein 2 like
View Full Product Details