Product Details

SNP ID
rs6523879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106617555 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTGCTTTAGATGTTACTATAAC[C/G]TGTGCAATTTCATAACTGATTAATG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CXorf57 PubMed Links

Gene Details

Gene
CXorf57
Gene Name
chromosome X open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184782.1 Intron NP_001171711.1
NM_018015.5 Intron NP_060485.4

View Full Product Details