Product Details

SNP ID
rs7991146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52020605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTATCTACCTCTTGATGGCGGTG[A/G]TAAAGTTTCATGATATCTGTACACA
Phenotype
MIM: 613666 MIM: 606882 MIM: 608969
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG11 PubMed Links

Gene Details

Gene
ALG11
Gene Name
ALG11, alpha-1,2-mannosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004127.2 Intron NP_001004127.2
Gene
ATP7B
Gene Name
ATPase copper transporting beta
There are no transcripts associated with this gene.

Gene
UTP14C
Gene Name
UTP14, small subunit processome component homolog C (S. cerevisiae)
There are no transcripts associated with this gene.

View Full Product Details