Product Details

SNP ID
rs7129768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:92972893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTCCTGTTTTAAAAGAACAAAA[A/G]ACTCTTTTCTTGACTTCATGTCCTC
Phenotype
MIM: 600804
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTNR1B PubMed Links

Gene Details

Gene
MTNR1B
Gene Name
melatonin receptor 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005959.3 Intron NP_005950.1
XM_011542839.2 Intron XP_011541141.1
XM_017017777.1 Intron XP_016873266.1

View Full Product Details