Product Details
- SNP ID
-
rs7893194
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:26218259 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCCTTGGGATGGCGGGTGGCCGA[C/T]GATCCAGGCGCTCGTGCGCTCACTG
- Phenotype
-
MIM: 138275
MIM: 606808
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GAD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78418694] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GAD2
- Gene Name
- glutamate decarboxylase 2
- Gene
- MYO3A
- Gene Name
- myosin IIIA
There are no transcripts associated with this gene.
View Full Product Details