Product Details
- SNP ID
-
rs7724957
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:179813626 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAAAACAATAATAAAAGCCAGG[C/G]ATGTTGGCATGTACCTGTAGTCCTA
- Phenotype
-
MIM: 604561
MIM: 601530
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MGAT4B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs57723546] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MGAT4B
- Gene Name
- mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B
There are no transcripts associated with this gene.
- Gene
- SQSTM1
- Gene Name
- sequestosome 1
View Full Product Details