Product Details

SNP ID
rs7066547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:116438597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGTTTATACCTTGTAGATCATT[A/G]GGGTAGACTCAATTAGAAAAAGGAG
Phenotype
MIM: 300444
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC6A14 PubMed Links

Gene Details

Gene
SLC6A14
Gene Name
solute carrier family 6 member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007231.4 Intron NP_009162.1

View Full Product Details