Product Details

SNP ID
rs6784645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172756486 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTTACATATCTGCTAAAATGATT[A/G]TGCTGATTTTCCTTTCTGTAAGTAT
Phenotype
MIM: 600586
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ECT2 PubMed Links

Gene Details

Gene
ECT2
Gene Name
epithelial cell transforming 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258315.1 Intron NP_001245244.1
NM_001258316.1 Intron NP_001245245.1
NM_018098.5 Intron NP_060568.3
XM_005247176.1 Intron XP_005247233.1
XM_006713523.3 Intron XP_006713586.1
XM_006713524.3 Intron XP_006713587.1
XM_011512514.2 Intron XP_011510816.1
XM_011512515.2 Intron XP_011510817.1
XM_011512516.1 Intron XP_011510818.1
XM_017005830.1 Intron XP_016861319.1
XM_017005831.1 Intron XP_016861320.1
XM_017005832.1 Intron XP_016861321.1
XM_017005833.1 Intron XP_016861322.1
XM_017005834.1 Intron XP_016861323.1
XM_017005835.1 Intron XP_016861324.1
XM_017005836.1 Intron XP_016861325.1
XM_017005837.1 Intron XP_016861326.1
XM_017005838.1 Intron XP_016861327.1

View Full Product Details