Product Details

SNP ID
rs6583602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142216259 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGAGCCTCAGCCATATGGCACTG[A/G]AATGAATGCTGCTGGGGAGATCTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00051 PubMed Links

Gene Details

Gene
LINC00051
Gene Name
long intergenic non-protein coding RNA 51
There are no transcripts associated with this gene.

Gene
TSNARE1
Gene Name
t-SNARE domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291931.1 Intron NP_001278860.1
NM_145003.4 Intron NP_659440.2
XM_011516912.1 Intron XP_011515214.1
XM_011516913.2 Intron XP_011515215.1
XM_011516914.1 Intron XP_011515216.1
XM_011516915.2 Intron XP_011515217.1
XM_011516916.1 Intron XP_011515218.1
XM_011516917.2 Intron XP_011515219.1
XM_011516918.1 Intron XP_011515220.1
XM_011516919.1 Intron XP_011515221.1
XM_011516920.1 Intron XP_011515222.1
XM_011516921.1 Intron XP_011515223.1
XM_011516922.1 Intron XP_011515224.1
XM_011516923.2 Intron XP_011515225.1
XM_011516924.1 Intron XP_011515226.1
XM_017013175.1 Intron XP_016868664.1
XM_017013176.1 Intron XP_016868665.1
XM_017013177.1 Intron XP_016868666.1
XM_017013178.1 Intron XP_016868667.1
XM_017013179.1 Intron XP_016868668.1
XM_017013180.1 Intron XP_016868669.1
XM_017013181.1 Intron XP_016868670.1

View Full Product Details