Product Details
- SNP ID
-
rs6520435
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:49607806 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGTGTATAATTATGACAATGGCTC[C/T]ATTTATTTTTAAAATAAGAGGAATT
- Phenotype
-
MIM: 300594
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GAGE1
PubMed Links
Gene Details
- Gene
- GAGE1
- Gene Name
- G antigen 1
View Full Product Details