Product Details

SNP ID
rs7675963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76220897 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCACCTGGGATCAGCTGGGACCA[C/G]AGGGGTTTCTTCATTCTCTCAGCGG
Phenotype
MIM: 602257
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM47E PubMed Links

Gene Details

Gene
FAM47E
Gene Name
family with sequence similarity 47 member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136570.2 Intron NP_001130042.1
NM_001242936.1 Intron NP_001229865.1
Gene
SCARB2
Gene Name
scavenger receptor class B member 2
There are no transcripts associated with this gene.

View Full Product Details