Product Details
- SNP ID
-
rs7583686
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:138672569 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTTGAGGGACAGGATCACTGTAT[A/T]TGATGCTGTTGCATGGAGTGAATAT
- Phenotype
-
MIM: 604635
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NXPH2
PubMed Links
Gene Details
- Gene
- NXPH2
- Gene Name
- neurexophilin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007226.2 |
|
Intron |
|
|
NP_009157.1 |
View Full Product Details