Product Details

SNP ID
rs7789409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123452248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAGGATGGAAGGTGAAAGCAGA[C/T]GATAAACTGCAAAGATTCAAATGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQUB PubMed Links

Gene Details

Gene
IQUB
Gene Name
IQ motif and ubiquitin domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282855.1 3460 UTR 3 NP_001269784.1
NM_001321293.1 3460 UTR 3 NP_001308222.1
NM_178827.4 3460 UTR 3 NP_849149.3
XM_005250161.3 3460 Intron XP_005250218.1
XM_005250162.4 3460 Intron XP_005250219.1
XM_011515833.2 3460 Intron XP_011514135.1
XM_011515834.2 3460 Intron XP_011514136.1
XM_017011771.1 3460 Intron XP_016867260.1
XM_017011772.1 3460 Intron XP_016867261.1

View Full Product Details