Product Details

SNP ID
rs7952008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:78443564 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGATCCAGTTCTGTCAGGCGCC[A/G]AGGCCCAGGGTCTTCACTCTGTTAC
Phenotype
MIM: 612803
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NARS2 PubMed Links

Gene Details

Gene
NARS2
Gene Name
asparaginyl-tRNA synthetase 2, mitochondrial (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243251.1 Intron NP_001230180.1
NM_024678.5 Intron NP_078954.4
XM_011545253.1 Intron XP_011543555.1
XM_017018302.1 Intron XP_016873791.1
XM_017018303.1 Intron XP_016873792.1
XM_017018304.1 Intron XP_016873793.1

View Full Product Details