Product Details

SNP ID
rs7189210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23840635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATACAAGCAGAGAGAGGATGT[C/T]GTCTCTGCCACTCTGGTAGGAGGTG
Phenotype
MIM: 176970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKCB PubMed Links

Gene Details

Gene
PRKCB
Gene Name
protein kinase C beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002738.6 Intron NP_002729.2
NM_212535.2 Intron NP_997700.1

View Full Product Details