Product Details

SNP ID
rs8080343
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30129740 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCATCAGTTTTAATCAGATAATG[C/T]CTTACTTCTGTAGATATAGTCTAGT
Phenotype
MIM: 616173
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NSRP1 PubMed Links

Gene Details

Gene
NSRP1
Gene Name
nuclear speckle splicing regulatory protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261467.1 Intron NP_001248396.1
NM_032141.3 Intron NP_115517.1
XM_011525345.2 Intron XP_011523647.1
XM_017025209.1 Intron XP_016880698.1
XM_017025210.1 Intron XP_016880699.1
XM_017025211.1 Intron XP_016880700.1
XM_017025212.1 Intron XP_016880701.1
XM_017025213.1 Intron XP_016880702.1
XM_017025214.1 Intron XP_016880703.1
XM_017025215.1 Intron XP_016880704.1

View Full Product Details