Product Details

SNP ID
rs7201010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:25237121 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCTTAGGATCAGAGCTTTCCTTGG[C/T]ACTGAGATCTAGATAAAAAAACCTT
Phenotype
MIM: 603750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AQP8 PubMed Links

Gene Details

Gene
AQP8
Gene Name
aquaporin 8
There are no transcripts associated with this gene.

Gene
ZKSCAN2
Gene Name
zinc finger with KRAB and SCAN domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012981.4 6006 UTR 3 NP_001012999.3
XM_017023200.1 6006 UTR 3 XP_016878689.1

View Full Product Details