Product Details

SNP ID
rs1805420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:147910788 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCCTTATCACAGCTGCAACTACT[C/G]ATTTACTTGTCTGACAATTTGATTT
Phenotype
MIM: 309550 MIM: 300805
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FMR1 PubMed Links

Gene Details

Gene
FMR1
Gene Name
fragile X mental retardation 1
There are no transcripts associated with this gene.

Gene
FMR1-AS1
Gene Name
FMR1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details