Product Details

SNP ID
rs7143400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52858173 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTCATGGCCCTAAGGAATGTGTG[A/C]CAAATTCAAGTTTATTATATCATAA
Phenotype
MIM: 607746
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FERMT2 PubMed Links

Gene Details

Gene
FERMT2
Gene Name
fermitin family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134999.1 2573 UTR 3 NP_001128471.1
NM_001135000.1 2573 Intron NP_001128472.1
NM_006832.2 2573 UTR 3 NP_006823.1
XM_005267285.2 2573 UTR 3 XP_005267342.1
XM_006720008.2 2573 UTR 3 XP_006720071.1
XM_006720009.2 2573 UTR 3 XP_006720072.1
XM_006720010.2 2573 UTR 3 XP_006720073.1

View Full Product Details