Product Details

SNP ID
rs7405631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82240314 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTCCCACCCAGCAGCTGAAGGGA[C/G]AAAGACGAGGAGGCAGGGAGCAGAC
Phenotype
MIM: 600864 MIM: 603877
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CSNK1D PubMed Links

Gene Details

Gene
CSNK1D
Gene Name
casein kinase 1 delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001893.4 Intron NP_001884.2
NM_139062.2 Intron NP_620693.1
XM_005256336.3 Intron XP_005256393.1
XM_005256337.4 Intron XP_005256394.1
XM_017024199.1 Intron XP_016879688.1
Gene
MIR6787
Gene Name
microRNA 6787
There are no transcripts associated with this gene.

Gene
SLC16A3
Gene Name
solute carrier family 16 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042422.2 Intron NP_001035887.1
NM_001042423.2 Intron NP_001035888.1
NM_001206950.1 Intron NP_001193879.1
NM_001206951.1 Intron NP_001193880.1
NM_001206952.1 Intron NP_001193881.1
NM_004207.3 Intron NP_004198.1
XM_011523607.1 Intron XP_011521909.1

View Full Product Details