Product Details

SNP ID
rs8111589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45531300 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCAGCTGGACTCACTGCTTCGTAT[C/T]GTGCCACATTTTCATTTTCTTTTAT
Phenotype
MIM: 606580 MIM: 601703
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OPA3 PubMed Links

Gene Details

Gene
OPA3
Gene Name
optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017989.2 Intron NP_001017989.2
NM_025136.3 Intron NP_079412.1
XM_006723403.3 Intron XP_006723466.1
Gene
VASP
Gene Name
vasodilator-stimulated phosphoprotein
There are no transcripts associated with this gene.

View Full Product Details