Product Details

SNP ID
rs8049970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72095419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAAAGTATGGAGAACATGGACGG[A/T]AGCCTTCTGCCCCATCTCTGTGCTG
Phenotype
MIM: 605584
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DHX38 PubMed Links

Gene Details

Gene
DHX38
Gene Name
DEAH-box helicase 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014003.3 Intron NP_054722.2
XM_005256269.1 Intron XP_005256326.1
XM_011523484.1 Intron XP_011521786.1
XM_011523485.1 Intron XP_011521787.1
XM_017023913.1 Intron XP_016879402.1
Gene
TXNL4B
Gene Name
thioredoxin like 4B
There are no transcripts associated with this gene.

View Full Product Details