Product Details

SNP ID
rs6776145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50278688 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAGCATTGTGTGCTAGCAGGTCA[C/T]GCAGTGGAGGCTGCTGTGTTTGCTG
Phenotype
MIM: 602725 MIM: 601281
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFRD2 PubMed Links

Gene Details

Gene
IFRD2
Gene Name
interferon related developmental regulator 2
There are no transcripts associated with this gene.

Gene
LSMEM2
Gene Name
leucine rich single-pass membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304385.1 379 Intron NP_001291314.1
NM_153215.2 379 Intron NP_694947.1
XM_006712979.3 379 UTR 5 XP_006713042.1
XM_006712980.3 379 UTR 5 XP_006713043.1
XM_011533370.2 379 Intron XP_011531672.1
Gene
MIR6872
Gene Name
microRNA 6872
There are no transcripts associated with this gene.

Gene
SEMA3B
Gene Name
semaphorin 3B
There are no transcripts associated with this gene.

View Full Product Details