Product Details

SNP ID
rs7953297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:63846732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCACTGCACCTGCCCTGCCCTG[A/T]GTCCAGGCTTTACCTCCTCCTGCAG
Phenotype
MIM: 606523
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SRGAP1 PubMed Links

Gene Details

Gene
SRGAP1
Gene Name
SLIT-ROBO Rho GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020762.2 Intron NP_065813.1
XM_005269042.3 Intron XP_005269099.1
XM_011538580.1 Intron XP_011536882.1
XM_011538581.2 Intron XP_011536883.1

View Full Product Details