Product Details

SNP ID
rs7725623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:102758794 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTGCCAGTATACAGCTATTGCT[C/T]TATGACTCAAGTAGTTATTTTAGTT
Phenotype
MIM: 170270
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAM PubMed Links

Gene Details

Gene
PAM
Gene Name
peptidylglycine alpha-amidating monooxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000919.3 Intron NP_000910.2
NM_001177306.1 Intron NP_001170777.1
NM_001319943.1 Intron NP_001306872.1
NM_138766.2 Intron NP_620121.1
NM_138821.2 Intron NP_620176.1
NM_138822.2 Intron NP_620177.1
XM_011543419.2 Intron XP_011541721.1
XM_017009495.1 Intron XP_016864984.1
XM_017009496.1 Intron XP_016864985.1
XM_017009497.1 Intron XP_016864986.1
XM_017009498.1 Intron XP_016864987.1
XM_017009499.1 Intron XP_016864988.1
XM_017009500.1 Intron XP_016864989.1
XM_017009501.1 Intron XP_016864990.1
XM_017009502.1 Intron XP_016864991.1
XM_017009503.1 Intron XP_016864992.1
XM_017009504.1 Intron XP_016864993.1
XM_017009505.1 Intron XP_016864994.1
XM_017009506.1 Intron XP_016864995.1
XM_017009507.1 Intron XP_016864996.1
XM_017009508.1 Intron XP_016864997.1
XM_017009509.1 Intron XP_016864998.1
XM_017009510.1 Intron XP_016864999.1
XM_017009511.1 Intron XP_016865000.1
XM_017009512.1 Intron XP_016865001.1
XM_017009513.1 Intron XP_016865002.1
XM_017009514.1 Intron XP_016865003.1
XM_017009515.1 Intron XP_016865004.1
XM_017009516.1 Intron XP_016865005.1

View Full Product Details