Product Details

SNP ID
rs121912591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44006157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTCGACTCCCTGGAGGTGACC[A/G]CGTCGCTGGCCAAGGCGCTGCGGCC
Phenotype
MIM: 608300 MIM: 600781
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAGS PubMed Links

Gene Details

Gene
NAGS
Gene Name
N-acetylglutamate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153006.2 725 Missense Mutation ACG,GCG T279A NP_694551.1
XM_011524438.1 725 Missense Mutation ACG,GCG T279A XP_011522740.1
XM_011524439.2 725 Missense Mutation ACG,GCG T113A XP_011522741.1
Gene
PYY
Gene Name
peptide YY
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004160.5 725 Intron NP_004151.3
XM_011525035.1 725 Intron XP_011523337.1
Gene
TMEM101
Gene Name
transmembrane protein 101
There are no transcripts associated with this gene.

View Full Product Details