Product Details

SNP ID
rs7198371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27203068 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTGCATACTTGGAGTCCACAGGAG[A/T]GTGTGCTCAAAACACAAAAGTCCCA
Phenotype
MIM: 611917
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KDM8 PubMed Links

Gene Details

Gene
KDM8
Gene Name
lysine demethylase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145348.1 Intron NP_001138820.1
NM_024773.2 Intron NP_079049.2
XM_017023676.1 Intron XP_016879165.1
XM_017023677.1 Intron XP_016879166.1
XM_017023678.1 Intron XP_016879167.1
XM_017023679.1 Intron XP_016879168.1
XM_017023680.1 Intron XP_016879169.1

View Full Product Details