Product Details

SNP ID
rs8111475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34733917 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGTTTTCTGGGAAGTGTAGTTTCT[C/T]GGTCCCGGCGCCATTGTCCTTAAAA
Phenotype
MIM: 606741
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF181 PubMed Links

Gene Details

Gene
ZNF181
Gene Name
zinc finger protein 181
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029997.3 215 Intron NP_001025168.2
NM_001145665.1 215 Intron NP_001139137.1
XM_005258849.2 215 Intron XP_005258906.1
XM_005258850.2 215 Intron XP_005258907.1
XM_005258851.3 215 Intron XP_005258908.1
XM_006723183.3 215 Intron XP_006723246.1
XM_006723184.3 215 Intron XP_006723247.1
XM_017026739.1 215 UTR 5 XP_016882228.1
XM_017026740.1 215 Intron XP_016882229.1

View Full Product Details