Product Details

SNP ID
rs5030320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:32388456 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGAGGTACTGGTTAGTTCTGATT[T/C]TTTCTTCAGCTGCTTGAAATGCATG
Phenotype
MIM: 607102
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
WT1 PubMed Links

Gene Details

Gene
WT1
Gene Name
Wilms tumor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000378.4 1747 UTR 3 NP_000369.3
NM_001198551.1 1747 UTR 3 NP_001185480.1
NM_001198552.1 1747 UTR 3 NP_001185481.1
NM_024424.3 1747 UTR 3 NP_077742.2
NM_024426.4 1747 UTR 3 NP_077744.3

View Full Product Details