Product Details

SNP ID
rs6062296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63660784 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTAGAGGGGCAGCCGGTGGCCAG[C/T]CTCACTTGGAAGGTCTTTGAACCTC
Phenotype
MIM: 608833 MIM: 608362
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RTEL1 PubMed Links

Gene Details

Gene
RTEL1
Gene Name
regulator of telomere elongation helicase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283009.1 Intron NP_001269938.1
NM_001283010.1 Intron NP_001269939.1
NM_016434.3 Intron NP_057518.1
NM_032957.4 Intron NP_116575.3
Gene
RTEL1-TNFRSF6B
Gene Name
RTEL1-TNFRSF6B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
STMN3
Gene Name
stathmin 3
There are no transcripts associated with this gene.

View Full Product Details