Product Details

SNP ID
rs8105115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11346702 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGGGGGCGGAGCCAGGATGATGA[C/T]CCCGCCCCCTCCCTAAATAATTCTC
Phenotype
MIM: 604350 MIM: 613771
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC159 PubMed Links

Gene Details

Gene
CCDC159
Gene Name
coiled-coil domain containing 159
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080503.2 Intron NP_001073972.2
XM_006722643.1 Intron XP_006722706.1
XM_011527678.2 Intron XP_011525980.2
XM_017026255.1 Intron XP_016881744.1
XM_017026256.1 Intron XP_016881745.1
XM_017026257.1 Intron XP_016881746.1
XM_017026258.1 Intron XP_016881747.1
XM_017026259.1 Intron XP_016881748.1
XM_017026260.1 Intron XP_016881749.1
XM_017026261.1 Intron XP_016881750.1
XM_017026262.1 Intron XP_016881751.1
XM_017026263.1 Intron XP_016881752.1
XM_017026264.1 Intron XP_016881753.1
Gene
PLPPR2
Gene Name
phospholipid phosphatase related 2
There are no transcripts associated with this gene.

Gene
RAB3D
Gene Name
RAB3D, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TMEM205
Gene Name
transmembrane protein 205
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145416.1 Intron NP_001138888.1
NM_001321112.1 Intron NP_001308041.1
NM_001321113.1 Intron NP_001308042.1
NM_001321114.1 Intron NP_001308043.1
NM_033408.3 Intron NP_212133.1
NM_198536.2 Intron NP_940938.1
XM_005259899.2 Intron XP_005259956.1
XM_017026769.1 Intron XP_016882258.1

View Full Product Details