Product Details

SNP ID
rs8002073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:100606682 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCCACCACCTGCCTCCCTGCAGT[G/T]GTTGGACTTCTGTAATGTTATGCAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMTC4 PubMed Links

Gene Details

Gene
TMTC4
Gene Name
transmembrane and tetratricopeptide repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079669.2 Intron NP_001073137.1
NM_001286453.1 Intron NP_001273382.1
NM_032813.3 Intron NP_116202.2
XM_011521121.2 Intron XP_011519423.1
XM_011521122.2 Intron XP_011519424.1
XM_011521123.2 Intron XP_011519425.1
XM_011521124.2 Intron XP_011519426.1
XM_011521125.1 Intron XP_011519427.1
XM_017020796.1 Intron XP_016876285.1
XM_017020797.1 Intron XP_016876286.1
XM_017020798.1 Intron XP_016876287.1
XM_017020799.1 Intron XP_016876288.1
XM_017020800.1 Intron XP_016876289.1

View Full Product Details