Product Details

SNP ID
rs7280687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30487336 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAGCTGCCACATCCACAGCCGT[A/G]GCCATAGCACATGCCACCAAAGCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP19-1 PubMed Links

Gene Details

Gene
KRTAP19-1
Gene Name
keratin associated protein 19-1
There are no transcripts associated with this gene.

Gene
KRTAP19-2
Gene Name
keratin associated protein 19-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181608.1 13 Missense Mutation CAC,TAC H5Y NP_853639.1
Gene
KRTAP19-3
Gene Name
keratin associated protein 19-3
There are no transcripts associated with this gene.

Gene
KRTAP19-4
Gene Name
keratin associated protein 19-4
There are no transcripts associated with this gene.

View Full Product Details