Product Details

SNP ID
rs7273305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2482350 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTGGACAGACAACGAGACAGAAA[A/G]TTGAGGATTCACTGGGGAGTGAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF343 PubMed Links

Gene Details

Gene
ZNF343
Gene Name
zinc finger protein 343
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282495.1 2963 UTR 3 NP_001269424.1
NM_001282496.1 2963 UTR 3 NP_001269425.1
NM_001282497.1 2963 UTR 3 NP_001269426.1
NM_001282498.1 2963 UTR 3 NP_001269427.1
NM_001282499.1 2963 Intron NP_001269428.1
NM_001321800.1 2963 UTR 3 NP_001308729.1
NM_001321801.1 2963 UTR 3 NP_001308730.1
NM_001321802.1 2963 UTR 3 NP_001308731.1
NM_001321803.1 2963 UTR 3 NP_001308732.1
NM_001321805.1 2963 UTR 3 NP_001308734.1
NM_024325.5 2963 UTR 3 NP_077301.4
XM_017028062.1 2963 UTR 3 XP_016883551.1

View Full Product Details